Learn More
Heidolph™ Viton™ Tubing
Product Code. 10689722 Shop All Heidolph Instruments Products
Click to view available options
Tubing Size:
0.8
1.7
7.9
Thickness (Metric):
1.6 mm
2.5 mm
Unit Size:
1m
Each
Product Code. 10689722
Brand: Heidolph™ 5255400000
Only null left
Add to basket
Add to basket
Use this tubing when you require excellent acid resistance at high temperatures. Heidolph™ Viton™ Tubing is smooth, biocompatible and inert.
  • Low gas permeability.
  • Resistant to solvents and corrosives at high temperatures.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Borosilicate Glass
Thickness (Metric) 1.6 mm
Sterility Non-sterile
Tubing Size 1.7
Maximum Pressure 0.7/1.7 bar