Learn More
Sartorius™ Vivacon™ Polycarbonate/Polypropylene Centrifugal Unit
Product Code. 11735785 Shop All Sartorius Products
Click to view available options
MWCO:
100 kDa
30 kDa
Unit Size:
Pack of 25
Product Code. 11735785
Brand: Sartorius™ VN01H21ETO
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
For Use With (Equipment) PCR
Material Polycarbonate, Polypropylene
MWCO 30 kDa
For Use With (Application) Concentration, Desalting, buffer exchange
Housing Material Polycarbonate, Polypropylene
Quantity 25/Pk.
Sample Type DNA
Type Vivacon 500 Centrifugal Unit
Volume (Metric) 0.1 to 0.5 mL