Learn More
Poulten Graf™ Volac™ Class A Borosilicate Glass Graduated Cylinders with Certificate
Product Code. 12328905
Click to view available options
Capacity (Metric):
5 mL
10 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
Product Code. 12328905
Brand: Poulten Graf™ R263/WAC/A/2
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Base Type Hexagonal
Calibration To Contain
Scale Color Blue
Shape Round
Certifications/Compliance ASTM E1272-02, EN ISO 4788
Class A
Type Graduated Cylinder
Material Borosilicate Glass
Capacity (Metric) 5 mL
Includes Single Certificate
Scale White Ceramic Glazed Ink Scale
Graduations 0.1 mL