Learn More
BRAND™ Volumetric Flask
Shop All Brand Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
Unit Size:
Each
Description
Specifications
Specifications
| Closure Material | Glass |
| Capacity (Metric) | 500 mL |
| Material | Borosilicate Glass (3.3) |
| Class | A |