missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vorinostat, Mikromol™
Description
Vorinostat
Specifications
Specifications
| Chemical Name or Material | Suberoylanilide Hydroxamic Acid |
| CAS | 149647-78-9 |
| Analyte or Component Names | Suberoylanilide Hydroxamic Acid |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C14 H20 N2 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | N1-Hydroxy-N8-phenyl-octanediamide, Octanediamide, N-hydroxy-N'-phenyl- (9CI), N-Hydroxy-N'-phenyloctanediamide, N1-Hydroxy-N8-phenyloctanediamide, SAHA, Suberoyl anilide bishydroxamide, Suberoylanilide hydroxamic acid, Vorinostat, Zolinza |
| InChI Formula | InChI=1S/C14H20N2O3/c17-13(15-12-8-4-3-5-9-12)10-6-1-2-7-11-14(18)16-19/h3-5,8-9,19H,1-2,6-7,10-11H2,(H,15,17)(H,16,18) |
| SMILES | ONC(=O)CCCCCCC(=O)Nc1ccccc1 |
| IUPAC Name | N'-hydroxy-N-phenyloctanediamide |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?