missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ VP-SFM AGT™ Medium
VP-SFM AGT™ Medium
Brand: Gibco™ 12559001
Additional Details : Weight : 2.40000kg
Description
VP-SFM AGT™ is a serum-free, ultra-low protein medium in an easy-to-use granular format. It is designed for the growth of VERO cells for virus production and for the growth of COS-7, MDCK, BHK-21 and Hep-2 cells for recombinant protein production.Advanced Granulation Technology = AGT
Specifications
Store at 2 to 8°C in the dark. | |
MDCK, VERO, HEp2, BHK-21, COS-7 | |
Suspension Cell Culture | |
VP Serum Free Medium (SFM) | |
Non-sterile | |
No Glutamine | |
Ropak Bucket | |
No antibiotics | |
Gibco™ | |
100 L | |
Wet Ice |
CO2 | |
Animal Origin-free, Low Protein, Serum-free | |
Powder (AGT) | |
Serum-free | |
Low Glucose, HEPES, Phenol Red, Sodium Pyruvate, Sodium Bicarbonate | |
Low | |
3.9 g/L | |
Calcium, Magnesium, Sodium Phosphate | |
Certified | |
100 L |
For Research Use Only. Not for use in diagnostic procedures.