Learn More
Velp Scientifica™ Rotor for OV5 Homogenizer
Velp Scientifica™ Rotor for OV5 Homogenizer can be reconfigured with a range of OV5 Homogenizer Shafts and Stators.
Brand:  Velp Scientifica™ A00000057
112.10 EUR valid until 2020-12-31
Use promo code "14720" to get your promotional price.
Additional Details : Poids : 0.03000kg
 View more versions of this product
Product Code. 15379544
Quantity Price
1 142.00€ / Each
10 130.00€ / Each
Expédition estimée
01-10-2020
Add to basket
Description
Description
  • Can be romoved and reconfigured
  • Interchangable
  • User-friendly
  • Strong and durable
Specifications
Specifications
Standard Rotor
1Each
30 mm
Dispenser
VSR3