missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VU 0409551
GREENER_CHOICE

Product Code. 18794888
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18794888

Brand: Tocris Bioscience™ 5693/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective mGlu5 positive allosteric modulator; brain penetrant and orally bioavailable

VU 0409551 is a selective mGlu5 positive allosteric modulator (EC50 = 260 nM). Selective for mGlu5 over other mGluR subtypes, as well as 66 other receptors and ion channels. Selectively potentiates mGlu5 signaling via Gαq. Exhibits wake-promoting, antipsychotic-like and cognition-enhancing effects in animal models. Brain penetrant and orally available.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material [6,7-Dihydro-2-(phenoxymethyl)oxazolo[5,4-c]pyridin-5(4H)-yl](fluorophenyl)methanone
CAS 1363281-27-9
Synonym JNJ 46778212
Quantity 50 mg
Target Group I mGluR Modulators
Molecular Formula C20H17FN2O3
Formula Weight 352.36
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.