missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VU 0409551

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
VU 0409551 is a selective mGlu5 positive allosteric modulator (EC50 = 260 nM). Selective for mGlu5 over other mGluR subtypes, as well as 66 other receptors and ion channels. Selectively potentiates mGlu5 signaling via Gαq. Exhibits wake-promoting, antipsychotic-like and cognition-enhancing effects in animal models. Brain penetrant and orally available.
Specifications
Specifications
| Chemical Name or Material | [6,7-Dihydro-2-(phenoxymethyl)oxazolo[5,4-c]pyridin-5(4H)-yl](fluorophenyl)methanone |
| CAS | 1363281-27-9 |
| Synonym | JNJ 46778212 |
| Quantity | 50 mg |
| Target | Group I mGluR Modulators |
| Molecular Formula | C20H17FN2O3 |
| Formula Weight | 352.36 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction