missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VU 6005649
GREENER_CHOICE

Product Code. p-7180142
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18709736

Brand: Tocris Bioscience™ 6893/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Positive allosteric modulators of mGlu7 and mGlu8 receptors. Also NK1 antagonist.

VU 6005649 is a positive allosteric modulator of mGlu7 and mGlu8 receptors (EC50 = 0.65 μM and 2.6 μM for mGlu7 and mGlu8, respectively). Displays >3.8 fold selectivity for mGlu7 and mGlu8 over other mGlu receptors (EC50 >10 μM for mGlu1,2,3,4,5,6). Also NK1 receptor antagonist (Ki= 650 nM, IC50 = 3.4 μM). Brain penetrant.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(2,3-Difluoro-4-methoxyphenyl)-2,5-dimethyl-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine
CAS 2137047-43-7
Quantity 10 mg
Molecular Formula C16H12F5N3O
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.