Learn More
Emma Safety Footwear Vulcanus Safety Shoes
Product Code. 15928711
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
Product Code. 15928711
Brand: Emma Safety Footwear MM53984848
Only null left
Add to basket
Add to basket
Welder's boot designed specifically for working with metal in extreme temperatures. Over-foot flap provides no entry for sparks or shards.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Velcro over-foot flap
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011
Color Black
Design Vulcanus
Liner Material Full grain Nappa /Hydro-Tec™ Sanitized Silver
Material Leather
Size 48