missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Wall Support accu-jet™ S
Accessory for use with accu-jet&trade- S pipette controller.
Brand: BRAND™ 26539
Additional Details : Weight : 0.01800kg
Description
- Accessory
- For use with accu-jet™ S
Specifications
Wall mount for accu-jet S | |
White | |
accu-jet S | |
Variable | |
Wall Support | |
21 g |
Autoclavable | |
Reusable | |
Polypropylene | |
Non-sterile | |
accu-jet™ S |