Learn More
Vitlab™ Wash Bottle Caps
Catalog Number 15762096
Click to view available options
Thread Size (Metric):
GL 45
GL 63
Formulated from Polypropylene Vitlab™ Wash Bottle Caps include precise spray jet and optimized medium backflow through the continuous and finely drawn tip of the spray tube.
  • Made of Polypropylene
  • Drawn-out tips
  • Precise spray
Material Polypropylene
No. per Pack 12/Pk
Thread Size (Metric) GL 45