Learn More
Rogo Sampaic™ Watch Glasses
Click to view available options
Capacity (Metric):
100 mL
1000 mL
150 mL
2 L
25 mL
250 mL
3 L
400 to 600 mL
50 mL
Unit Size:
Pack of 10
Pack of 20
Description
- Available in a range of diameters
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Autoclavable | Autoclavable |
| Capacity (Metric) | 25 mL |
| Material | Borosilicate Glass |
| Type | Watch Glass |
| For Use With (Application) | Beaker |
| Temperature Range (Metric) | 500 °C |
| Diameter (Metric) | 40 mm |