Learn More
Rogo Sampaic™ Watch Glasses
Product Code. p-8000408
Click to view available options
Capacity (Metric):
100 mL
1000 mL
150 mL
2 L
25 mL
250 mL
3 L
400 to 600 mL
50 mL
Unit Size:
Pack of 10
Pack of 20
Product Code. 11702724
Brand: Rogo Sampaic™ KBN013
Only null left
Add to basket
Edge
Add to basket
Excellent for liquid evaporation, weighing solids, heating small amounts of substances and as beaker covers. Rogo Sampaic™ Watch Glasses are manufactured from durable clear borosilicate glass.
  • Available in a range of diameters
  • Corrosion-free and resistant to thermal shock
Specifications
Autoclavable Autoclavable
Capacity (Metric) 100 mL
Material Borosilicate Glass
Type Watch Glass
For Use With (Application) Beaker
Temperature Range (Metric) 500 °C
Diameter (Metric) 60 mm