Learn More
Gilac™ Water Erasable Marker Pens, Black
Product Code. 17928971
Click to view available options
Color:
Black
Unit Size:
Each
Product Code. 17928971
Brand: Gilac™ G020520
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Black Water Erasable Marker Pens are specially designed to write on plastic containers, crates, and buckets.
  • GILAC water erasable marker pens are specially designed to write on plastic containers, crates, and buckets.
  • IML label (label overmolded inside the plastic material rendering it permanent and rewritable) facilitates the HACCP approach thanks to a traceability management.
  • Pack of 10 marker pens.
  • Black water erasable ink.
  • Fine tip: 0.5 mm
  • Non-food ink.
Specifications
Tip Style Fine
Product Type Washable Marker
Color Black
Quantity 10 Pk.
Thickness 0.5 mm