missing translation for 'onlineSavingsMsg'
Learn More

Water, for Karl Fischer Analysis, Solution In Methanol (1Ml = 5.0mg/H2O), Fisher Chemical™

Catalog Number 10598050
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber glass bottle
Plastic Coated Glass bottle
This item is not returnable. View return policy
11332
This item is not returnable. View return policy
11332
Color Undesignated
Quantity 500 mL
Boiling Point 64.7°C
Packaging Plastic Coated Glass bottle
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Water solution Karl Fischer Laboratory Reagent
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'