missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ Water Testing

Product Code. 11943757
Click to view available options
Format:
Tube
Unit Size:
Pack of 25
This item is not returnable. View return policy

Product Code. 11943757

Brand: Palintest™ PL416

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

These water testing tablets are ideal for quick and reliable testing in the field.

  • Individually wrapped in aluminum
  • Total phosphorus tube tests
  • Measure phosphorus for photometers
  • X24 phosphate tests
  • Net weight: 0.80000kg

Specifications

Format Tube
Quantity 25 Pack
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : sodium metabisulphite, adipic acid, ammonium molybdate, boric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.