Learn More
Palintest™ Water Testing
Click to view available options
Format:
Tube
Unit Size:
Pack of 25
Description
- Individually wrapped in aluminum
- Total phosphorus tube tests
- Measure phosphorus for photometers
- X24 phosphate tests
- Net weight: 0.80000kg
Specifications
Specifications
| Format | Tube |
| Quantity | 25 Pack |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- EUH031-Contact with acids liberates toxic gas.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : sodium metabisulphite, adipic acid, ammonium molybdate, boric acid