Learn More
Tempshield™ Waterproof Cryo-Grip™ Gloves
Description
- Grip palm for improved dexterity handling small objects
- Thin, seamless 100% waterproof liner provides protection from spills and splashes
- High level of thermal protection when exposed to cryogenic fluids
- For ultra-cold applications down to -196°C
- CE Category III: EN 511, EN 388, EN 420
Specifications
Specifications
| Certifications/Compliance (Europe) | CE Cat. III, EN 511, EN 388, EN 420 |
| Color | Pink |
| Product Type | Cryogenic Gloves |
| Size | Medium |
| Style | Wrist |
| Temperature (Metric) Max | -196°C |
| For Use With (Application) | Handling of smaller objects or samples stored or hardened in cryogenic liquids, dispensing/transferring cryogenic liquids, any open cryogenic containers with the chance of exposure to liquids or splashes |
| Series | Tempshield™ Cyro-Protection™ |