Learn More
Zwiebel™ Weights Set
Product Code. 11850962
Click to view available options
Weight (Metric):
1110 g (Total)
210 g (Total)
6110 g (Total)
Description:
E2 Series
F1 Series
Unit Size:
Each
Product Code. 11850962
Brand: Zwiebel™ PBI56847
Only null left
Add to basket
Add to basket
Manufactured to the highest standard. Zwiebel™ Weights Set are precise measuring tools.
  • Stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability/In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Product Type Calibration Weight
Certifications/Compliance Cofrac calibration certificate
Description F1 Series
Material Antimagnetic Stainless Steel
Case Material Polypropylene
Weight (Metric) 1110 g (Total)
Shape Cylindrical
Includes Case includes gripper