missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Welding Cut Gloves
Click to view available options
Size:
7
8
9
Description
- Suitable for MIG and ARC welding in dry or light oily conditions with cut hazards.
- Water repellent, heatproof cowhide grain leather back.
- Water-repellent, heatproof velvet cowhide palm.
- Water repellent, heatproof split leather cuff to protect the forearm.
- U.S. assembly.
- Gunn cut, winged thumb with cord, Crispin type.
- Elasticated at the back of the hand.
- Kevlar™ cotton lining for comfort and thermal insulation. Outstanding comfort and flexibility.
Specifications
Back Material | Cowgrain leather |
Certifications/Compliance | CE CATEGORY II Intermediary risks, NF EN 420-03 EEC Basic standard, NF EN 388-03 Mechanical risks, NF EN 407-04 Thermical risks, EN 12477-02 Type A Welding risks, EC Category PPE 2, EC Certificate Number 0075/797/162/10/12/0763, EC Attestation Number 0075/797/162/10/12/0763 |
Classification | Dexterity: 5 |
Cuff Material | Split leather |
Cut Style | Gunn |
Cut Resistance Rating | 2 |
Material | Cowhide Leather |
Product Type | Crispin Welding Gloves (Type A) |
Size | 7 |
Thickness (Metric) | 0.8/1.1 mm (Back), 1.1/1.3 mm (Cuff) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'