Learn More
Cole-Parmer™ Werther PC3/50/37 Ultra-Quiet Oilless Air Compressor
Product Code. 15804158 Shop All Cole-Parmer Products
Change view
Click to view available options
Flow Rate:
7.39 CFM, 207 L/min.
Unit Size:
Each
Product Code. Flow Rate unitSize
15804158 7.39 CFM, 207 L/min. Each
1 options
Product Code. 15804158
Brand: Cole-Parmer™ 0706765
Only null left
Add to basket
Add to basket
Werther PC3/50/37 Ultra-Quiet Oilless Air Compressor Tanks are coated with powder-based paint to resist harsh environments and make them corrosion-resistant. Pressure switch operates between 90 and 120 psi.
  • Ultra-quiet operation as low as 30 dB(A)
  • Provide dry, clean air
  • Comes with a storage tank for instant high-pressure, pulse-free air
Specifications
Horsepower 2 hp
Shipping Weight (English) 154 lb.
Shipping Weight (Metric) 70 kg
Amperage 10 amp
Dimensions (L x W x H) 63.5 x 55.8 x 73.66 cm (25 x 22 x 29 in.)
Description Ultra-Quiet Oilless Air Compressor
Material 314 Stainless Steel, 316 Stainless Steel, Aluminum
Noise Level 78 dB
Temperature (English) Max. 150°F
Temperature (Metric) Max 66°C
Flow Rate 7.39 CFM, 207 L/min.
Frequency 60 Hz
Pump Type Vacuum System
Maximum Pressure 101 psi
Voltage 220 V