Learn More
ZVG Zellstoff Vertrieb™ Polytex™ Wet Wipes
Description
- Solvent-resistant and washable
- 100% Polypropylene fabric, blue
- Featured on a roller or stack
- Robust and lint-free
Specifications
Specifications
| Product Type | Wet Wipes |
| Length (Metric) | 42 cm |
| Width (Metric) | 40 cm |
| Material | Polypropylene |
| Absorbancy | High |
| Color | Blue |
| Dimensions (L x W) | 40 x 32 cm |
| For Use With (Application) | For numerous applications |
| Packaging Quantity | 36 Cartons |