Learn More
Cytiva Whatman™ Anotop™ 10 Sterile Syringe Filters
Hold-up volume: <30μL (10mm size), <200μL (25mm)
Brand: Cytiva 6809-1122
Additional Details : Weight : 0.10800kg
Description
Hold-up volume: <30μL (10mm size), <200μL (25mm)
Similar to Anotop filters, but include binder-free, glass microfiber prefilter with Anotop™ inorganic membrane in a polypropylene housing.
Glass Microfiber Prefilter
- Increases throughput
- Maintains membrane performance
- Reduces need for centrifuging before filtering, sequential filtering, or refiltering
- Permits use of 0.1μm pore size Anotop Plus filters to remove mycoplasma contaminants from tissue culture
- Hold-up volume: <30μL (10mm size), <200μL (25mm)
- Pressure limit, 7kg/cm (100psi)
Specifications
Anotop 10 mm Sterile Syringe Filters, 0.2 μm (50 pcs) | |
Anapore | |
Sterile | |
HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water | |
Polypropylene (PP) | |
Male luer | |
50 Pieces | |
45°C | |
Syringe Filter |
Single-use | |
0.2 μm | |
0.78 cm2 | |
100 mL | |
Female luer lock | |
6.9 bar (100 psi) | |
113°F | |
Polypropylene | |
10 mm |