Learn More
Cytiva Whatman™ AS 600 Series Multiple Vacuum Filtration Apparatus
Click to view available options
Volume (Metric):
100 mL
500 mL
Includes:
Stainless Steel support screen filter, 6 place manifold
Stainless Steel support screen filter, 6 place manifold, rapid closure clamp
Unit Size:
Each
Description
AS 600 Series
The apparatus can be autoclaved and sterilized by dry heat at up to 180°C. Suitable only for vacuum operation. If flushing tubes are used, do not exceed 1.3 bar (300 mbar over-pressure).
The apparatus can be autoclaved and sterilized by dry heat at up to 180°C. Suitable only for vacuum operation. If flushing tubes are used, do not exceed 1.3 bar (300 mbar over-pressure).
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Stainless Steel |
| Includes | Stainless Steel support screen filter, 6 place manifold |
| Series | AS600/3 Series |
| Type | Vacuum Filtration System |
| Volume (Metric) | 500 mL |