Learn More
Cytiva Whatman Cyclopore Polycarbonate Membrane
Product Code. 17149880
Change view
Click to view available options
Unit Size:
Pack of 100
Pack of 25
Pore Size:
0.2 μm
0.4 μm
0.5 μm
5 μm
Dimensions (L x W):
209.55 x 292.1 mm
25 x 80 mm
Diameter (Metric):
293 mm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize Pore Size Dimensions (L x W) Diameter (Metric)
17149880 Pack of 25 - - -
17199880 Pack of 25 0.5 μm 209.55 x 292.1 mm -
17189880 Pack of 100 5 μm - -
17806797 Pack of 100 5 μm 25 x 80 mm -
17179880 Pack of 25 - - -
17159880 Pack of 100 0.2 μm - 293 mm
17169880 Pack of 25 0.4 μm - 293 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17149880 Supplier Cytiva Supplier No. 10417050
Only null left
Add to basket
Add to basket
Featuring sharp cut-off and reproducible microfiltration characteristics of track-etched membrane filters
  • Low affinity for stains improves visibility under microscope
  • Low protein binding minimizes sample loss of proteinaceous samples
  • Narrow pore size distribution and high pore density support rapid surface capture of analyte
  • Negligible absorption and adsorption of filtrate; nonhygroscopic
  • No particle shedding enables clean filtrate
  • Biologically inert
TRUSTED_SUSTAINABILITY
Specifications
Material Polycarbonate
For Use With (Application) Epifluorescence Microscopy, Environmental Analysis, Cell Biology, EPA Testing, Fuel Testing, Bioassays, Parasitology, Air Analysis, Water Microbiology
Quantity 25 Pk.
Type Membrane Circle