Learn More
Cytiva Whatman™ Drying Rack for 903 Protein Saver Cards
Air-dries blood specimens in a suspended horizontal position. Whatman™ Drying Rack for 903 Protein Saver Card is easily assembled and can be affixed to a wall or counter top with optional hook-and-loop stickers.
37.55€ - 93.20€
Specifications
Product Type | Drying Rack |
---|---|
For Use With (Equipment) | 903 Protein Saver Card |
Quantity | 10 Pack |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
11909134
|
Cytiva
10539521 |
Hook-and-Loop Mounting Sticker |
93.20€
Pack of 10 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
11359394
|
Cytiva
10537173 |
Mounting Sticker not included |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Descripción
Air-dries blood specimens in a suspended horizontal position. Whatman™ Drying Rack for 903 Protein Saver Card is easily assembled and can be affixed to a wall or counter top with optional hook-and-loop stickers.
- For use with neonatal human diagnostic testing cards
- CE Marked
- Accommodate multiple collection forms at one time
Especificaciones
Drying Rack | |
903 Protein Saver Card | |
10 Pack |