Learn More
Cytiva Whatman™ Funnels for Microbiological Filtration System
Product Code. 11309284
Click to view available options
:
PP funnel; 350mL
Product Code. 11309284
Brand: Cytiva 10445866
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Autoclavable plastic funnel for the MBS I system.
  • Simple to use.
  • Safe sealing mechanism.
  • Shorter preparation time.
  • High reproducibility.
  • Funnels can be autoclaved up to 50 times.
  • Large funnel capacity for foaming liquids.
  • Easy to validate.
  • Risk of cross-contamination is minimized.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Microbiological filtration system (MBS I)
Volume (Metric) 350 mL