Learn More
QIAGEN QIAcard FTA Elute Indicating Micro
Product Code. 12445432
Click to view available options
Quantity:
100 Pk.
25 Pk.
Unit Size:
Each
Pack of 25
Product Code. 12445432
Brand: QIAGEN WB120412
For collection, stabilization, processing, transport and archiving of biological samples. 1 sample area per card.
  • FTA Elute technology for convenient water based elution of nucleic acids.
  • QIAcard Indicating FTA Cards include a pink dye that turns white when a colorless sample, e.g. buccal cells or saliva is applied.
  • Stabilization and protection of nucleic acids at room temperature with for spot volumes of 125 μL.
  • Cell lysis on contact, denatures proteins and protects DNA from degradation.
Specifications
Content And Storage Room Temperature
Description Collect, process, transport, and store colorless biological samples, such as buccal or saliva samples. With FTA Elute, proteins remain tightly bound while DNA is eluted from the matrix.
For Use With (Application) Multiplex PCR, Sequencing after PCR amplification, SNP analysis, STR analysis, HLA typing, Whole genome amplification, Quantitative PCR, Biobanking, Pharmacogenomics, Genotyping, Genetic identification, Transgenic detection
Includes Inert dye
Product Type Indicating FTA Elute Micro Card
Quantity 25 Pk.
Sample Volume 40 μL
Test Time 15 to 30 min
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501-Dispose of contents/container to .