Learn More
Cytiva Whatman™ ME 25 Plain Mixed Cellulose Ester Membranes
Product Code. 11345314
Click to view available options
Diameter (Metric):
25 mm
50 mm
90 mm
100 mm
110 mm
142 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 25
Pack of 50
Product Code. 11345314
Brand: Cytiva 10401672
Facilitates the counting process and minimizes eye fatigue with color contrast between the surface and the colonies. Whatman™ ME 25 Plain Mixed Cellulose Ester Membranes are characterized by a smooth and more uniform surface.
  • Excellent contrast for easier particle detection.
  • Offers a high degree of internal surface area for greater adsorption of product.
  • Higher dirt loading capacity.
  • Biologically inert with good thermal stability.
  • No surfactants to contaminate samples.
  • Uniform microporous structure of membrane gives high flow rates.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Bubble Point 2.8 bar
Color White
Material Cellulose Acetate, Cellulose Nitrate
Pore Size 0.45 μm
Sterility Sterile
Flow Rate 12.5 sec./100 mL/12.5 cm2
Includes Interleaving paper
Membrane Type ME25
Quantity 100/Pk.
Thickness (Metric) 145 μm
Diameter (Metric) 50 mm
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement