Learn More
Cytiva Whatman™ Mini-UniPrep™ Amber Syringeless Filters
Produced with the same colorant used in pharmaceutical containers. Whatman™ Mini-UniPrep™ Amber Syringeless Filters, Slit Septa provide an easier way to remove particulates from HPLC/UHPLC samples in one-third the time as compared to traditional syringe filters.
Brand: Cytiva US203APUNYL
Additional Details : Weight : 0.28000kg
Description
Whatman Mini-UniPrep™ Amber Syringeless Filters quickly and easily remove particulates from light-sensitive samples being prepared for HPLC/UHPLC analysis, while meeting USP specifications for light resistance.
- Pre-assembled all-in-one filtration unit eliminates the need for syringe filter, syringe, vial and septum, reducing costs by up to 40%
- Compatible with most major autosamplers for high-throughput analysis
- Fewer consumables required
- Plunger contains a nylon membrane at one end and a pre-attached cap/slit septum at the other
- Can be used either manually or with a compressor unit
- Multi-sample compressor processes up to 6 samples at a time, further improving sample processing time and reducing hand stress
- Designed to fit into any autosampler accommodating 12 x 32mm vials -OR-
Slit septum can be easily, repeatedly pierced with needle and sample drawn off for manual injection into an analyzer
Specifications
Amber | |
Single-use | |
PTFE-coated Silicone Rubber Septa | |
0.45 μm | |
Polypropylene (PP) | |
Split septa | |
0.6 bar / 8 psi (to compress) | |
Syringeless Filter |
Equivalent in size to 12 x 32mm vials | |
Autosampler, Compressor | |
Nylon | |
Routine HPLC/UHPLC analysis; Composite assays; Content uniformity; Protein precipitation; Solubility testing; Dissolution testing; Sample filtration; Light sensitive samples; Ion chromatography | |
Vial with incorporated filtration membrane, plunger and cap/split septa | |
100/Pk. | |
50°C | |
0.4 mL |