Learn More
Cytiva Whatman™ Polyamide Membranes
Product Code. 11375404
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
0.5 μm
Diameter (Metric):
25 mm
47 mm
50 mm
Unit Size:
Pack of 100
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Diameter (Metric) unitSize
11375404 0.2 μm 25 mm Pack of 100
11395404 0.2 μm 50 mm Pack of 100
11345174 0.5 μm 25 mm Pack of 100
11305414 0.45 μm 47 mm Pack of 100
11358944 0.5 μm 50 mm Pack of 100
11385404 0.2 μm 47 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11375404 Supplier Cytiva Supplier No. 10414006
Only null left
Add to basket
Add to basket
Universal filter made from pure polyamide for clarification and sterile filtration
  • Mechanicall strong
  • Excellent wet/dry strength
  • Hydrophilic for aqueous and organic solutions
  • For up to 135°C
TRUSTED_SUSTAINABILITY
Specifications
Material Polyamide
Pore Size 0.2 μm
Hydrophobicity Hydrophilic
Thickness (Metric) 110 μm
Diameter (Metric) 25 mm