Learn More
Cytiva Whatman™ Polycap Disposable Capsules, 75 AS
Product Code. 11364614
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Inlet Style:
Female National Pipe Thread
Stepped Barb
Stepped Barb with filling bell
Outlet Style:
Female National Pipe Thread
Stepped Barb
Stepped Barb with filling bell
Sterility:
Non-sterile
Sterile
Unit Size:
Each
Pack of 5
Product Code. 11364614
Brand: Cytiva 67057502
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Polycap 75 AS filters for filtration of aqueous solutions
  • Sterile and nonpyrogenic
  • Integrity testable
  • With inlet vent
  • Filter Media: Low-extractable, hydrophilic nylon membrane and glass microfiber prefilter
  • Filling bell model (09-744-61) minimizes contamination when filtering into bottle or flask
Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material (Membrane) Nylon
Pore Size 0.2 μm
Sterility Sterile
Filtration Area 820 cm2
For Use With (Application) Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.
Housing Material Polypropylene
Inlet Style Stepped Barb
Outlet Style Stepped Barb