Learn More
Cytiva Whatman™ Reeve Angel 802 Prepleated Qualitative Circles
Product Code. 11761598
Click to view available options
Diameter (Metric):
12.5 cm
15 cm
18.5 cm
24 cm
32 cm
38.5 cm
Unit Size:
Pack of 100
Product Code. 11761598
Brand: Cytiva 5802125
Only null left
Add to basket
Add to basket
Coarse retention
  • Folded for fast filtration of coarse particles and gelatinous precipitates
  • Qualitative Grade; nominal retention 30-35μm; 0.26mm thick Pleating and creped surface increase effective area, improving both filtering speed and extending filter life
  • Wet strengthened†
TRUSTED_SUSTAINABILITY
Specifications
Description 802 12.5 cm 100/pk
Format Disc
Particle Retention 15 μm
Pore Size 30 to 35 μm
For Use With (Application) A prepleated filter for use with a conical filter funnel, offers fast filtration and high loading capacity for analysis involving coarse particles or gelatinous precipitates.
Grade 802
Porosity Coarse
Quantity 100/Pk.
Thickness (Metric) 0.26 mm
Dimensions (L x W) 12.50 cm
Diameter (Metric) 12.5 cm