Learn More
Cytiva Whatman™ Regenerated Cellulose Membranes: Grade RC 60
Product Code. 11313365
Click to view available options
Diameter (Metric):
47 mm
50 mm
Unit Size:
Pack of 100
Product Code. 11313365
Brand: Cytiva 10410014
Only null left
Add to basket
Add to basket
Consists of pure cellulose without any wetting agents
  • Spontaneously wetting, very good wet strength
  • Extremely chemically resistant; suitable for aqueous and organic media
  • Hydrophilic
  • Mechanically stable
  • Sterilizable by all methods
TRUSTED_SUSTAINABILITY
Specifications
Material Regenerated Cellulose
Pore Size 1.0 μm
Hydrophobicity Hydrophilic
Thickness (Metric) 70 μm
Diameter (Metric) 50 mm