missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Black Phenolic Open Top Cap with Gray Butyl Septa and Flange
Shop All Wheaton Science Products Products
Click to view available options
No. per Case:
100 Case
1000 Case
Unit Size:
Pack of 100
Description
Replacement Black Phenolic Screw Caps for Media Bottles
- Open Top Cap With Gray Chlorobutyl / 50 Septa and Flange
- Autoclavable
Specifications
Specifications
| Color | Black |
| Includes | Gray Butyl septa and flange |
| Material | Phenolic Resin |
| No. per Case | 100 Case |
| Neck Finish | 33-430 |
| Certifications/Compliance | ISO 9001 |
| Product Type | Open Top Cap |
| For Use With (Equipment) | Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction