Learn More
DWK Life Sciences Wheaton™ BOD Bottles Replacement Robotic Stopper
Product Code. 11708879 Shop All Wheaton Science Products Products
Click to view available options
Shape:
Pennyhead
Robotic
Unit Size:
Pack of 12
Product Code. 11708879
Brand: DWK Life Sciences 227670
Only null left
Add to basket
Add to basket
Compatible with 60 and 300mL BOD bottles
  • Glass, closure-style stopper
  • Unlined
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 60 and 300 mL BOD Bottle
Material Glass
Shape Robotic