Learn More
DWK Life Sciences Wheaton™ Center Disc Tear-Out Aluminum Seals, Unlined
Tear-out seals remove completely allowing content to pour from bottle. Wheaton™ Center Disc Tear-Our Aluminum Seals, Unlined are available in a wide range of sizes and colors.
Brand: DWK Life Sciences 224183-07
126.35 EUR valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 0.63500kg
Description
- Good barrier properties
- Color selection includes natural, red, blue and green
- Chemically resistant to alcohol and acetone, among others
Specifications
Green | |
1000/Cs. | |
20 mm |
Aluminum | |
Center Disc Tear-out Cap |