Learn More
DWK Life Sciences Wheaton™ Glass Sample Vials in Lab File without Caps
Shop All Wheaton Science Products Products
Click to view available options
Volume (Metric):
2 mL
4 mL
8 mL
12 mL
16 mL
20 mL
24 mL
25 mL
30 mL
40 mL
Color:
Amber
Clear
Packaging Type:
2 x Shrink-wrapped Packs
Partitioned Tray
Unit Size:
Pack of 200
Description
Partitioned trays provide an easy way to inventory samples or to store empty vials.
- Autoclavable
- Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
- Amber vials made from borosilicate glass that protects light-sensitive products
Order Info
Caps available separately
Compliance
Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements. Amber vials conform to USP Type 1 requirements for light transmission.
Specifications
Specifications
| Color | Clear |
| Diameter (Metric) Outer | 12 mm |
| Material | Borosilicate Glass |
| Volume (Metric) | 2 mL |
| Packaging Type | Partitioned Tray |
| Thread Style | Screw |
| Volume (English) | 0.5 dr. |
| Autoclavable | Autoclavable |
| Closure Type | Screw Cap |
| Height (Metric) | 35 mm |
| Show More |