Learn More
DWK Life Sciences Wheaton™ Graduated Valumetric™ Bottles: With White PP Cap With Polyvinyl Liner
Product Code. p-4902645 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
Quantity:
144/Pk.
24/Pk.
288/Pk.
432/pk.
48/Pk.
96/Pk.
Unit Size:
Pack of 144
Pack of 24
Pack of 288
Pack of 432
Pack of 48
Pack of 96
Product Code. 10021382
Brand: DWK Life Sciences W216966
Only null left
Add to basket
Add to basket
For storage and samples of solids, dry powdered materials and liquids
  • Ideal for use as sample bottles
  • Useful for dry powders, soil samples, and specimen storage
  • Made from Wheaton 800 glass, which meets USP Type III standards
  • Graduated in mL and oz.
Autoclavable black phenolic cap with rubber liner.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Clear
Diameter (English) 1.3 in.
Material Glass
Quantity 48/Pk.
Closure Color White
Height (Metric) 70 mm
Height (English) 2.7 in.
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Size 33-400 mm
Diameter (Metric) 34 mm
Closure Type Screw Cap