Learn More
DWK Life Sciences Wheaton™ Impellers for Celstir™ Spinner Flasks
Product Code. p-123068 Shop All Wheaton Science Products Products
Click to view available options
For Use With (Equipment):
1000mL Celstir Spinner Flask
125mL Celstir Spinner Flask
250mL Celstir Spinner Flask
3000mL Celstir Spinner Flask
500mL Celstir Spinner Flask
50mL Celstir Spinner Flask
Unit Size:
Each
Product Code. 10168611
Brand: DWK Life Sciences 356904
Only null left
Add to basket
Add to basket
Replacement for Double Sidearm Celstir and Jacketed Double Sidearm Celstir.
  • This non-breakable stainless steel shaft, for replacing the glass shaft included with Celstir™ flasks, may be retrofitted to the existing Celstir flasks.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 1000mL Celstir Spinner Flask
Includes Top cap with liners, glass rod, magnet holder, magnet and stirring paddle
Type Impeller