missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Low-Density Extended Controlled Dropper Tips (15mm)
Shop All Wheaton Science Products Products
Click to view available options
Color:
Blue
Gray
Green
Natural
Orange
Pink
Purple
Red
White
Yellow
Material:
LDPE
Polyethylene
No. per Case:
100 Cs.
1000 Cs.
Unit Size:
Pack of 100
Pack of 1000
Description
- Extended controlled dropper tip dispenses one drop at a time
- Average drop size is 44μL ±6μL
- Case of 1000
Specifications
Specifications
| Color | Green |
| Material | Polyethylene |
| No. per Case | 1000 Cs. |
| Certifications/Compliance | ISO 9001 |
| Drop Size | 41 μL |
| Product Type | Dropper Tip |
| For Use With (Equipment) | Plastic dropping bottle |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion