Learn More
DWK Life Sciences Wheaton™ Micro Tissue Grinders
Product Code. 11531305 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity:
0.003 oz., 0.1mL
0.006 oz., 0.2mL
0.02 oz., 0.75mL
Length (English):
3.46 in.
4.33 in.
4.52 in.
Tube Diameter (English) Outside:
0.15 in.
0.27 in.
0.31 in.
Unit Size:
Pack of 2
Pack of 25
Product Code. Capacity Length (English) Tube Diameter (English) Outside unitSize
11531305 0.003 oz., 0.1mL 4.33 in. 0.15 in. Pack of 2
10449651 0.006 oz., 0.2mL 4.52 in. 0.27 in. Pack of 2
11551305 0.02 oz., 0.75mL 3.46 in. 0.31 in. Pack of 25
3 options
Product Code. 11531305
Brand: DWK Life Sciences 357844
Only null left
Add to basket
Add to basket
Used in extremely precise work when delicate hand operation is required
  • Made from low extractable borosilicate glass
Potter-Elvehjem Micro Tissue Grinder
  • Reservoir and pouring lip provide easy decanting of micro sample
  • Unit can be autoclaved / sterilized to prevent cross contamination between samples
  • Pestle clearance range: 0.1 – 0.15mm
Micro Tissue Grinder with PTFE Spacer
  • PTFE spacer aligns the pestle and reduces aerosoling of the product
  • Includes a finely ground glass pestle and tube
  • Autoclavable
Disposable Micro Tissue Grinder
  • Disposable to eliminate cross contamination and useful for radioactive samples
  • Small grinder area allows for high sample recovery
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Capacity (English) 0.003 oz.
Capacity (Metric) 0.1 mL
Material Borosilicate Glass
Product Type Micro Disposable Tissue Grinder
For Use With (Application) Used in extremely precise work when delicate hand operation is required
Length (English) 4.33 in.
Length (Metric) 110mm
Length (English) Tube 2.5 in.
Length (Metric) Tube 65mm
Tube Diameter (English) Outside 0.15 in.
Tube Diameter (Metric) Outside 4mm
Capacity 0.003 oz., 0.1mL