missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Microlink Caps

Product Code. p-76192 Shop All Wheaton Science Products Products
Click to view available options
Neck Finish:
33-430
38-430
Color:
Black
White
Product Type:
Open Top Screw Cap
Screw Cap
No. per Case:
24/Cs.
50/Cs.
Unit Size:
Pack of 24
Pack of 50
This item is not returnable. View return policy

Product Code. 11501235

Brand: DWK Life Sciences 240106

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Open top caps are ideal for storing and containing volatile solvents, sensitive compounds and corrosive chemicals

  • Polypropylene/silicone/PTFE liner is ultrasonically welded to each cap to help eliminate possibility of toxicity
  • Autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Polypropylene
No. per Case 50/Cs.
Liner Material Polypropylene/Silicone
Neck Finish 33-430
Certifications/Compliance ISO 9001
Product Type Screw Cap
For Use With (Equipment) Bottle
Autoclavable Autoclavable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.