Learn More
DWK Life Sciences Wheaton™ Open Top Screw Caps with Silicone Liner
Catalog Number 12643375
Click to view available options
Neck Finish:
13-425
15-425
Color:
Black
White
No. per Case:
1000 Cs.
250 Cs.
Open top screw caps have a bonded PTFE faced silicone liner that provides access with a syringe.
  • Can withstand autoclaving / sterilization
Closure Liner PTFE/Silicone
For Use With (Equipment) Glass bottle or vial
Product Type Open Top Screw Cap
Color Black
Material Phenolic Resin
No. per Case 1000 Cs.
Neck Finish 15-425
Certifications/Compliance ISO 9001