Learn More
DWK Life Sciences Wheaton™ Peroxide Cured Silicone Tubing For General Lab Use
Product Code. p-4383797 Shop All Wheaton Science Products Products
Click to view available options
Diameter (Metric) Inner:
2 mm
3 mm
6 mm
8 mm
Overall Length (Metric):
150 cm
760 cm
Unit Size:
Each
Product Code. 12411531
Brand: DWK Life Sciences 374308
Only null left
Add to basket
Add to basket
Assembly consists of 1.5m (5 ft.) section of silicone tubing with a wall thickness of 2.3mm, a borosilicate glass sinker, and a polypropylene delivery nozzle.
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Inner 31/100 in.
Diameter (Metric) Inner 8 mm
Material Silicone
Overall Length (English) 25 ft.
Overall Length (Metric) 760 cm
Thickness (English) 0.09 in.
Thickness (Metric) 2.3 mm
For Use With (Application) Peristaltic pump
Product Type Tubing