missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps for S/T Bottles
Shop All Wheaton Science Products Products
Click to view available options
Color:
Black
White
Product Type:
Cap
Open Top Cap
Unit Size:
Pack of 300
Description
- Unlined
- Open- and closed-top caps
- 20-400 screw thread
- Autoclavable
Specifications
Specifications
| Color | Black |
| Material | Polypropylene |
| No. per Case | 300/Cs. |
| Neck Finish | 20-400 |
| Certifications/Compliance | ISO 9001 |
| Product Type | Cap |
| For Use With (Equipment) | Screw neck diagnostic bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction