Learn More
Wheaton™ Shelf
Shop All Wheaton Science Products Products
Click to view available options
For Use With (Equipment):
Roll-in incubator
Unit Size:
Each
Description
For use with Wheaton™ Roll-In Incubators
Specifications
Specifications
| For Use With (Equipment) | Roll-in incubator |
| Type | Shelf |
| Dimensions (L x W) | 24 x 34.4 in. (61 x 87.4 cm) |
| Length (English) | 24 in. |
| Length (Metric) | 61 cm |
| Width (English) | 34.4 in. |
| Width (Metric) | 87.4 cm |
| Depth (English) | 24 in. |
| Depth (Metric) | 61 cm |