missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ White Polypropylene Screw Caps for Glass Bottles and Vials
Shop All Wheaton Science Products Products
Click to view available options
Neck Finish:
15-425
18-400
24-400
Liner Material:
PTFE/Polyethylene
PTFE/Silicone
Polyethylene Foam
Polyvinyl
No. per Case:
144/Cs.
200/Cs.
Unit Size:
Pack of 144
Pack of 200
Description
- Available with different liners (PTFE/silicone, foamed polyethylene, poly-vinyl, PTFE faced foamed polyethylene)
- With choice of open or closed top
Caps with PTFE/silicone liner:
- Provides heat resistance
- Glass filled
- Suited for EPA samples
- Chemical resistant
- Tight seal
- With moisture barrier
- Suited for mild acids, alkalis, solvents, alcohols, oils and aqueous products
- Chemical resistant
- Provides compressibility and sealing properties
- Suited for high purity chemicals, strong acids and solvents
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| No. per Case | 144/Cs. |
| Liner Material | Polyvinyl |
| Neck Finish | 18-400 |
| Certifications/Compliance | ISO 9001 |
| For Use With (Equipment) | Glass bottle or vial |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction